GC columns, J&W HP-101
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
19091Y-012 19091Y-015 19091Y-102
HEWL19091Y-012EA 724 EUR
HEWL19091Y-012 HEWL19091Y-015 HEWL19091Y-102
GC columns, J&W HP-101
Kolommen voor chromatografie
HP-101 is a 100% dimethylpolysiloxane phase for established methods developed on non-bonded or cross-linked columns.
  • Each Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provide you utmost confidence in qualitative and quantitative results
  • For details about the QC test conditions including the test probe mixture and the resulting chromatogram, please refer to the Agilent J&W GC Column Performance Summary shipped with each GC column
It is not bonded or cross-linked, and is therefore useful for established methods that do not require the column to have cross-linked or bonded polymer phases.
Opgelet: Because HP-101 columns are not bonded or cross-linked, solvent rinsing is not recommended.