InterPlay Adenoviral TAP Systems
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240214 240216
AGLS240214EA 527 EUR
AGLS240214 AGLS240216
InterPlay Adenoviral TAP Systems
Vectoren, Plasmiden en Bibliotheken
The InterPlay Adenoviral TAP System combines our unique tandem affinity purification (TAP) system with our exclusive adenoviral gene delivery system, the AdEasy Adenoviral Vector System, for enhanced gene delivery to a broader range of mammalian cells, higher protein yields, and improved purification and analysis of endogenous interacting protein partners.
  • Purify interacting proteins from a broader range of mammalian cells
  • Recombinant adenoviruses can infect either dividing or non-dividing cells
Opgelet: For research use only. Not for use in diagnostic procedures.