ISIS 3
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5023-1517 5023-1522 5023-1523 5023-1524 5023-1525 5023-1527 5043-0054 5043-0055 G8411-67111 G8411-67115 G8411-80115 G8411-80119 G8411-80121
HEWL5023-1517EA 31 EUR
HEWL5023-1517 HEWL5023-1522 HEWL5023-1523 HEWL5023-1524 HEWL5023-1525 HEWL5023-1527 HEWL5043-0054 HEWL5043-0055 HEWLG8411-67111 HEWLG8411-67115 HEWLG8411-80115 HEWLG8411-80119 HEWLG8411-80121
ISIS 3
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The ISIS 3 dramatically increases the productivity of both the Agilent 7800 ICP-MS and the Agilent 7900 ICP-MS while also allowing high matrix samples to be run for extended periods of time.
  • Constant flow nebulisation eliminates stabilisation after high-speed uptake
  • Reduces the amount of sample reaching the instrument, reducing cleaning and maintenance requirements
  • Provides the highest possible throughput with discrete sampling
With its fast uptake piston pump and close-coupled 7-port valve located close to the nebuliser, the ISIS 3 enables reduced sample uptake and rinse times with discrete sampling (DS). The ISIS 3 delivers unmatched productivity for high sample matrix analysis in high throughput laboratories.