Speciation columns and supplies
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G1820-65541 G3154-65002 G3288-80000 G3154-65001 G3268-80001
HEWLG1820-65541EA 984 EUR
HEWLG1820-65541 HEWLG3154-65002 HEWLG3288-80000 HEWLG3154-65001 HEWLG3268-80001
Speciation columns and supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
High quality Speciation Columns used as replacements and accessories in the ICP-MS systems portfolio.
  • Excellent peak separation and reproducibility for the most common speciation methods
  • Excellent reproducibility of both the retention time (RT) and concentration
  • Helps users determine quantity and chemical species with confidence
  • Greater productivity – avoid the need for re-analysis of samples
  • The broadest range of phases and column configurations
A range of chromium speciation columns, arsenic guard columns and high precision speciation columns for the testing of arsenic in drinking water or in urine/food. Ensure samples and supplies are safe, accurate and compliant with regular testing in accordance with local legislation. To maintain system performance, ensure that consumables are periodically replaced in accordance with recommended maintenance schedules.