QuikChange II
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
200555 200521 200522 200523 200524 200555
STRA200555EA 0 EUR
STRA200555 AGLS200521 AGLS200522 AGLS200523 AGLS200524 AGLS200555
QuikChange II
Reagentia voor nucleïnezuren Nucleic Acid Modification and Labelling
These QuikChange II kits provide improved fidelity, while maintaining greater than 80% mutation efficiency for single site mutagenesis.
  • Includes PfuUltra High-fidelity DNA polymerase to minimise unwanted errors
  • Complete kit provides all reagents necessary to support mutagenesis (at single sites)
Used for large constructs, and for use with electroporation competent cells.
Opgelet: For research use only. Not for use in diagnostic procedures.