Graphite platforms for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0422 8003-0960
HEWL8003-0422EA 822 EUR
HEWL8003-0422 HEWL8003-0960
Graphite platforms for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These graphite platforms and tubes for PerkinElmer graphite furnace AA systems are quality tested to ensure every batch passes our demanding performance specifications for contamination, sensitivity, precision, electrical resistance, and lifetime.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
L'vov platforms isolate the sample from the tube walls to allow indirect heating of theMore Product Information