Capillaries for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0509 8003-0510
HEWL8003-0509EA 205 EUR
HEWL8003-0509 HEWL8003-0510
Capillaries for PerkinElmer
Slangen Chromatography Tubing
These capillaries are equivalent to the PerkinElmer ICP-MS versions.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system