Glycan mapping HPLC columns, AdvanceBio
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
820750-905 821725-905 821725-906 858700-913 859700-913
HEWL820750-905EA 838 EUR
HEWL820750-905 HEWL821725-905 HEWL821725-906 HEWL858700-913 HEWL859700-913
Glycan mapping HPLC columns, AdvanceBio
Kolommen voor chromatografie
AdvanceBio Glycan Mapping Columns for fast, reproducible, high resolution glycan characterisation.
  • Speed: Obtain glycan maps in less than 10 minutes, superior results – in 40% less time than the competition
  • High-resolution: Characterise both labeled and unlabeled glycans, and identify any glycosylation changes that occur due to process variables
  • Flexibility: 2,7 µm superficially porous particles for high resolution with less backpressure. For UHPLC analysis 1,8 µm particles are an ideal choice
  • Reproducibility: QA tested with a 2-AB labeled IgG N-linked glycan sample to ensure consistent quality and performance
AdvanceBio Glycan Mapping columns leverage technology that optimises results for MS and fluorescence detection. Choose from two UHPLC configurations: 2,7 µm superficially porous, for high resolution and lower backpressure, or 1,8 µm for highest resolution.
Order AdvanceBio Glycan Mapping columns with a selection of standards for performance testing and retention mapping of labeled and unlabeled glycans. Our AdvanceBio N-glycan sample preparation kits provide all the components needed for the sample prep workflow with a comprehensive procedure to simplify the process.