AAV helper-free system
Leverancier: AGILENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
240109 240071 240073 240074 240075 240109
STRA240109EA 0 EUR
STRA240109 AGLS240071 AGLS240073 AGLS240074 AGLS240075 AGLS240109
AAV helper-free system
Vectoren, Plasmiden en Bibliotheken
The AAV Helper-Free System eliminates the requirement for wild-type adenovirus co-infection from both AAV vector production and AAV stock titering steps, making this system entirely helper virus-free and safer. This system works with a broad range of hosts and infects both dividing and non-dividing cells.
  • Stable gene expression
  • High titer virus
  • Lower toxicity vs. non-viral methods of gene delivery
Opgelet: For Research Use Only. Not for Use in Diagnostic Procedures.