GC columns, J&W GS-Alumina
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
115-3532 115-3552
HEWL115-3532EA 956 EUR
HEWL115-3532 HEWL115-3552
GC columns, J&W GS-Alumina
Kolommen voor chromatografie
GS-Alumina is the most polar alumina phase and is an excellent general use column for light hydrocarbon analysis.
  • Most polar Alumina phase
  • Aluminum oxide with proprietary deactivation
  • Excellent general use column for light hydrocarbon analysis: C1 to C8 hydrocarbon isomers
  • Separates C1 to C4 saturated and unsaturated hydrocarbons
  • Best for resolving cyclopropane from propylene
Alumina columns have a tendency to adsorb water and CO₂ which, over time, result in changes in retention time. Agilent uses an advanced, proprietary deactivation process that allows for rapid regeneration, and so fully moisture saturated GS-Alumina columns regenerate completely within 7 hours or less at 200 °C.