Stabiliser for sensitive biomolecules, SeramunStab® STB
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIL3119-100ML
BIL3119-250ML
APOSBIL3119-100MLEA
132
EUR
APOSBIL3119-100ML
APOSBIL3119-250ML
Stabiliser for sensitive biomolecules, SeramunStab® STB
Koppelings- en labelingssystemen voor eiwitten
Liquid product colourless to slightly yellow, freshly solutions opaleszent, odour weakly pH 7,3 to 7,5 boiling point 104 °C. Stabiliser for sensitive biomolecules like proteohormones or membrane proteins.