SeramunBlock
Leverancier: Apollo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIL3122-100ML
BIL3122-250ML
APOSBIL3122-100MLEA
0
EUR
APOSBIL3122-100ML
APOSBIL3122-250ML
SeramunBlock
Koppelings- en labelingssystemen voor eiwitten
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.