BM-1 orchid medium
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMB141A-10X1L PMB141C-50L PMB141N-10L
APOSPMB141A-10X1LEA 36.8 EUR
APOSPMB141A-10X1L APOSPMB141C-50L APOSPMB141N-10L
BM-1 orchid medium
Celcultuurmedia
Contains macro/micronutrients, vitamins and plantgrowth regulators required for orchid culturePlant tissue culture testedFull specification given in appendix.
  • MFCD07370630