Burner chamber assemblies for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0944 8003-0945
HEWL8003-0944EA 933 EUR
HEWL8003-0944 HEWL8003-0945
Burner chamber assemblies for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Agilent burner chamber assemblies for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
Instruction guides and how-to videos are available to ensure you get the best performance from your PerkinElmer system when using our parts and supplies.