Murashige and skoog modified basal salts (no nitrogen)
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMM531A-10X1L PMM531B-10L PMM531C-50L
APOSPMM531A-10X1LEA 73.5 EUR
APOSPMM531A-10X1L APOSPMM531B-10L APOSPMM531C-50L
Murashige and skoog modified basal salts (no nitrogen)
Celcultuurmedia
Contains macro/micronutrients as described by Murashige and Skoog, 1962. Modified by eliminating NH₄NO₃ and KNO₃ content. Plant tissue culture tested. Full specification given in appendix.
  • MFCD00240976