Murashige and skoog modified basal salts (½ nitrogen)
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMM561-10X1L PMM561-10L
APOSPMM561-10X1LEA 0 EUR
APOSPMM561-10X1L APOSPMM561-10L
Murashige and skoog modified basal salts (½ nitrogen)
Celcultuurmedia
Contains macro/micronutrients as described byMurashige and Skoog, 1962. Modified by reducing NH₄NO₃ and KNO₃ contentPlant tissue culture testedFull specification given in appendix.
  • MFCD00240976