H1 oat medium
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMO612A-10X1L PMO612B-10L
APOSPMO612A-10X1LEA 0 EUR
APOSPMO612A-10X1L APOSPMO612B-10L
H1 oat medium
Celcultuurmedia
Contains agar and sucrose. Contains the macro- and micronutrients and supplements required for the symbiotic culture of orchids.
  • Plant tissue culture tested