Serological pipettes
Leverancier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F110121 F110123 F110125 F110127 F110129 F110122 F110124 F110126 F110128 F110130 F110131
GILSF110121EA 136.4 EUR
GILSF110121 GILSF110123 GILSF110125 GILSF110127 GILSF110129 GILSF110122 GILSF110124 GILSF110126 GILSF110128 GILSF110130 GILSF110131
Serological pipettes
Pipetten Serologische pipetten
The Gilson disposable plastic serological pipettes are perfect for use with MACROMAN® to help you in your daily cell-culture, molecular biology and microbiology applications. Ideal for scientists who need to easily and quickly transfer liquid aliquots from one container to another with precision.
  • USP Class VI polystyrene and are free of contaminants that can alter your experiment
  • Gamma-sterilised, non-pyrogenic, and certified free of detectable RNase/DNase
  • Polyester fiber plug, offering a high level of protection against over-pipetting and contamination by liquids and aerosols
  • Colour-coded rings for easy size identification
  • Dark ascending and descending graduations so you can easily read the aspirated and dispensed volumes, they also have negative graduations for extra capacity
Available in models ranging from 1 to 50 ml, these serological pipettes are reco mmended for use with MACROMAN® but can also be used with any pipette controller. They enable precise and convenient dispensing of different kinds of liquids for a variety of tasks. Two kinds of packaging for various applications and precise sampling.