Spray chambers and brackets
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
110868000 G8010-60256 G8010-80036 3710035300 3710037800 G8010-60271 G8010-68011 G8010-80042 G8010-80043 6910006000
VARI110868000EA 187 EUR
VARI110868000 HEWLG8010-60256 HEWLG8010-80036 HEWL3710035300 HEWL3710037800 HEWLG8010-60271 HEWLG8010-68011 HEWLG8010-80042 HEWLG8010-80043 HEWL6910006000
Spray chambers and brackets
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.