Hyperskimmer cones for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0579 8003-0580 8003-0581 8003-0584
HEWL8003-0579EA 333 EUR
HEWL8003-0579 HEWL8003-0580 HEWL8003-0581 HEWL8003-0584
Hyperskimmer cones for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These hyperskimmer cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage.