Big traps, BMT/BOT series
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BMT-2 BOT-2 BOT-4
HEWLBMT-2EA 436 EUR
HEWLBMT-2 HEWLBOT-2 HEWLBOT-4
Big traps, BMT/BOT series
Filters Gasfilters
Big moisture/oxygen traps ideal for bulk purification applications or where several instruments are plumbed from a single gas source.
  • One piece of heavy-walled aluminum tube eliminates potential leaks
  • Pressure: Up to 17,23 bar
  • 750 cm³ gas purifiers
Equipped with sintered stainless steel frits to prevent particulate contamination.