J&W DB-1 GC columns, megabore
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
124-1032 124-1034 125-1005 125-100B 125-1011 125-1017 125-101J 125-101K 125-1025 125-102J 125-1035E 125-1037 125-1039 125-103B 125-103J 125-103K 125-1055 125-1064 125-1065 125-106J 125-10H5 125-10HB 125-1055E
HEWL124-1032EA 883 EUR
HEWL124-1032 HEWL124-1034 HEWL125-1005 HEWL125-100B HEWL125-1011 HEWL125-1017 HEWL125-101J HEWL125-101K HEWL125-1025 HEWL125-102J HEWL125-1035E HEWL125-1037 HEWL125-1039 HEWL125-103B HEWL125-103J HEWL125-103K HEWL125-1055 HEWL125-1064 HEWL125-1065 HEWL125-106J HEWL125-10H5 HEWL125-10HB HEWL125-1055E
J&W DB-1 GC columns, megabore
Kolommen voor chromatografie
DB-1 is a reliable, cross-linked and non-polar column with low bleed and a high temperature limit, delivering superb performance for general purpose applications.
  • 100% Dimethylpolysiloxane
  • Non-polar and low bleed with a high temperature limit
  • Wide range of applications
  • Bonded, cross-linked and solvent rinsable
  • Equivalent to USP Phase G2