J&W DB-200 GC columns
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
122-2032 122-2033 123-2032 123-2033 125-2032
HEWL122-2032EA 772 EUR
HEWL122-2032 HEWL122-2033 HEWL123-2032 HEWL123-2033 HEWL125-2032
J&W DB-200 GC columns
Kolommen voor chromatografie
DB-200 is a mid-polarity column ideal for difficult-to-separate positional isomers.
  • (35% Trifluoropropyl)-methylpolysiloxane
  • Temperature limit - 300/320 °C
  • Mid polarity - more polar than DB-1701 or DB-17
  • Ideal for difficult-to-separate positional isomers
  • Unique interactions with compounds containing nitro, halogen and carbonyl groups
J&W DB-200 is a (35% trifluoropropyl)-methylpolysiloxane column. This mid-polarity column is ideal for difficult-to-separate positional isomers. It has unique interactions with compounds containing nitro, halogen and carbonyl groups.