Skimmer cones for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0572 8003-0574 8003-0576 8003-0578
HEWL8003-0572EA 536 EUR
HEWL8003-0572 HEWL8003-0574 HEWL8003-0576 HEWL8003-0578
Skimmer cones for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These skimmer and sample cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.