Pistons and seals
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G4220-60015 G5611-60016 G1376-60015 5067-5678
HEWLG4220-60015EA 390 EUR
HEWLG4220-60015 HEWLG5611-60016 HEWLG1376-60015 HEWL5067-5678
Pistons and seals
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These pistons are made from a high purity, mono-crystalline sapphire for maximum durability.
  • Consistent, reproducible results
Although ceramic pistons can be manufactured at a lower cost, ceramic is a sintered, polycrystalline material, which can cause undesirable variations during the production process. Agilent seals are designed to fit snugly around our pistons, and are capable of adapting to a wide range of flow rates and pressures. The combination of our piston and seal has undergone extensive testing under temperature stress, with all commonMore Product Information