Vacuum degassers
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5042-8922 5067-5388
HEWL5042-8922EA 139 EUR
HEWL5042-8922 HEWL5067-5388
Vacuum degassers
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
A vacuum degasser is recommended for maximum sensitivity in the low UV wavelength range; high injection precision; high retention time reproducibility; and flow rates below 0,5 ml/min.
  • To generally clean the vacuum degasser tubing, flush the system with isopropanol
  • Flush the degasser with water after using buffers
Speed solvent changes by drawing solvent through the degasser and tubing with syringe adapter kit.