Polypropylene vials
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5190-2242 5190-3155
HEWL5190-2242EA 43 EUR
HEWL5190-2242 HEWL5190-3155
Polypropylene vials
flesjes
These vials are appropriate for pH sensitive samples, sodium, and heavy metal analyses.
  • Manufactured from virgin PP
  • Chemically resistant
Wide opening glass/plastic vials have a glass flanged insert molded to the inside, so sample only comes in contact with high quality glass insert and septum.
Certificaten: Meets requirements of 21 CFR 177.1520.