Capillaries for Shimadzu LC systems
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8001-0818 8001-0821 8001-0822 8001-0824
HEWL8001-0818EA 74 EUR
HEWL8001-0818 HEWL8001-0821 HEWL8001-0822 HEWL8001-0824
Capillaries for Shimadzu LC systems
Slangen Chromatography Tubing
Stainless steel capillaries and tubing for Shimadzu HPLC Systems. Ideal for most standard applications.
  • Made of ultra-clean stainless steel tubing
  • Ideal for most standard applications, and the best choice for reliable high-pressure sealing
  • Smooth inner surface for the lowest backpressure
  • Precision cutting for burr-free, square-cut ends without inner-diameter distortion
  • Both ends accept standard fittings
  • Predefined lengths for specific path locations to avoid peak broadening
  • Individually tested to ensure reproducibility
Used within combination with theMore Product Information