Electron multipliers
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5190-0108 5190-0123 G8400-60258
HEWL5190-0108EA 2605 EUR
HEWL5190-0108 HEWL5190-0123 HEWLG8400-60258
Electron multipliers
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The electron multiplier converts incident ions into an amplified electric current used to record measurements of the ion signal in ICP-MS Systems.
As the voltage in the system increases the amplification factor (or gain) increases. In general the electron multiplier can measure a count of 1.6E10. The system should be replaced when the total ion count reaches more than 3.2E11 and the sensitivity is less than 4000 counts with 10 ppb Y (even when applying detector voltage above 2700 V).