GC columns, J&W DB-502.2
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
123-1464 125-14A4
HEWL123-1464EA 1420 EUR
HEWL123-1464 HEWL125-14A4
GC columns, J&W DB-502.2
Kolommen voor chromatografie
Agilent JW DB-502.2 columns deliver excellent peak shapes for EPA method 502.2 and are available in 105 m lengths for volatiles analysis.
  • Excellent peak shape
  • Bonded and cross-linked
  • Solvent rinsible
Agilent J&W GC column is tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency to provides utmost confidence in qualitative and quantitative results. Precision-engineered DB-502.2 columns also have an operating temperature range of 0 to 260/280 °C.