Bond elut C18 OH plates
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7542703C 7541601C
VARI7542703CEA 464 EUR
VARI7542703C VARI7541601C
Bond elut C18 OH plates
Microplaten Platen voor vastefase-extractie
Bond Elut C18 OH is unendcapped, offering tightly controlled silanol activity to support fractionation. C18 OH delivers enhanced retention of polar analytes.
  • Silanol activity permits metabolite fractionation
  • Tight QC tolerances deliver batch-to-batch reproducibility
  • 150 Å pore size expands retention window to higher molecular weight compounds
It is a nonendcapped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.