Burners for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0417 8003-0418 8003-0419 8003-0858
HEWL8003-0417EA 1803 EUR
HEWL8003-0417 HEWL8003-0418 HEWL8003-0419 HEWL8003-0858
Burners for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
Agilent burner chamber assemblies for PerkinElmer AA systems are designed to provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
The 5 cm nitrous oxide burner head is required for nitrous oxide-acetylene operation, but can also be used with air-acetylene or rotated to provide reduced sensitivity. A 5 cm air-acetylene burner head isMore Product Information
Order Now
Specificaties
Environmentally Preferable