PSD spares and supplies
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6610025900 9910091200 9910115100 1510146800
VARI6610025900EA 254 EUR
VARI6610025900 VARI9910091200 VARI9910115100 HEWL1510146800
PSD spares and supplies
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
The Programmable Sample Dispensers (PSD) are high-capacity autosamplers that are used with Graphite Furnace AA.
  • Plug and play mounting with an ultrastable mounting and position lock, eliminating re-alignment
  • Provides capacity for up to up to 50 samples in disposable 2 ml microvials, plus up to five central vessels for blank, bulk standard, and chemical modifiers
  • Can automatically perform sample preparation processes for GFAAS, eliminating time-consuming manual sample preparation
  • Provides flexible dispensing options that are commonly used in GFAAS, including hot inject, multiple injection, and pre- or postaddition of modifiers