GC columns, J&W DB-5ht
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
122-5711 122-5731 123-5711 123-5731 123-5711E 123-5731E 122-5711E 122-5731LTM 123-5701 123-5701LTM
HEWL122-5711EA 543 EUR
HEWL122-5711 HEWL122-5731 HEWL123-5711 HEWL123-5731 HEWL123-5711E HEWL123-5731E HEWL122-5711E HEWL122-5731LTM HEWL123-5701 HEWL123-5701LTM
GC columns, J&W DB-5ht
Kolommen voor chromatografie
DB-5ht is a nonpolar column specially processed for an extended temperature limit of 400 °C.
  • (5%-Phenyl)-methylpolysiloxane
  • Non-polar
  • Specially processed for extended temperature limit of 400 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
J&W DB-5ht nonpolar columns are manufactured from (5% phenyl) methylpolysiloxane. They deliver high levels of performance in a variety of analytical processes. High temperature limits of –60 to +400 °C result from precision engineeredMore Product Information