HPLC columns, ZORBAX® Eclipse, XDB-C8
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
7995108-344 7995108-585 7995108-595 930990-906 931975-936 933975-936 935967-906 960967-906 963967-906 966967-906 971700-906 973700-936 975700-936 990967-206 990967-306 993700-906 961600-906 961967-906 963600-906 965600-906 961753-906 961967-306 922975-906 932967-906 972700-906 974700-906 927700-906K 927975-306K 927975-906K 928700-906K 928975-306K 928975-906K 935967-906K 961967-906K 963967-906K 990967-906K 993967-906K
HEWL7995108-344EA 716 EUR
HEWL7995108-344 HEWL7995108-585 HEWL7995108-595 HEWL930990-906 HEWL931975-936 HEWL933975-936 HEWL935967-906 HEWL960967-906 HEWL963967-906 HEWL966967-906 HEWL971700-906 HEWL973700-936 HEWL975700-936 HEWL990967-206 HEWL990967-306 HEWL993700-906 HEWL961600-906 HEWL961967-906 HEWL963600-906 HEWL965600-906 HEWL961753-906 HEWL961967-306 HEWL922975-906 HEWL932967-906 HEWL972700-906 HEWL974700-906 HEWL927700-906K HEWL927975-306K HEWL927975-906K HEWL928700-906K HEWL928975-306K HEWL928975-906K HEWL935967-906K HEWL961967-906K HEWL963967-906K HEWL990967-906K HEWL993967-906K
HPLC columns, ZORBAX® Eclipse, XDB-C8
Kolommen voor chromatografie
Agilent ZORBAX® Eclipse XDB-C8 columns provide four bonded phase choices for method development optimisation.
  • Long lifetime
  • Four selectivity choices for method development optimisation
  • Good peak shape for basic, acidic and neutral compounds
  • High performance over a wide pH range pH 2 to 9
  • Particle sizes available in 1,8, 3,5, 5 and 7 μm for high speed, high resolution, analytical and preparative scale separations
These columns provide good peak shape over a wide pH range (2 to 9) for additional method development flexibility with one family of columns. Eclipse XDB columns can be used for method development at low pH (2 to 3) and the same column can be used for method development in the mid pH (6 to 8) region. In the mid pH region, residual silanols are more active and tailing interactions are more likely. To overcome these interactions, Eclipse XDB columns are extra densely bonded and double end capped through a proprietary process to cover as many active silanols as possible. The result is superior peak shape of basic compounds from pH 2 to 9.