Microbial air samplers, Mas-100 Iso Mh®
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAS-100
1.19166.0001EA 1600 EUR
1.19166.0001 1.17174.0001 1.17309.0001 1.19154.0001 1.17312.0001 1.17290.0001 1.17292.0001 1.19380.0001 1.17291.0001 1.17310.0001
Microbial air samplers, Mas-100 Iso Mh®
Monsternemers Luchtmonsternametoestellen
Highest collection efficiency and safety in isolators and rabs with up to four sampling points.
  • Precise, reproducible measurements and outstanding user flexibility
  • Excellent collection efficiency
  • Fully automatic calibration
The microbial air sampler MAS-100 Iso MH is used for microbial monitoring of air in isolators and RABS. It can be configured with one to four sampling heads. The sampling heads with the nutrient plates are located in the containment, while the control unit with electronics and all moving parts is located outside the enclosure and aspirates the air through a pipe/tube system. The system is designed for maximum protection of the product and the environment and can be controlled fully automatically via various data interfaces.
Technical Specifications:
One to four sampling heads per control unit
Air flow rate 100 standard liters per minute (SLPM)
Accuracy of air flow ±2,5%
Physical cut off diameter d50 = 1,1 µm with standard perforated lid 300×0,6 mm
Sampling head made of stainless steel EN 1.4435 (X2CrNiMo18-14-3) according to BN2 (electropolished pharmaceutical grade) with surface roughness RA <0,8 µm
In-line decontamination of the sampling head and the air aspiration channel
IP 65 protection as a standard
Length of base unit 38 cm, width 23 cm, height 12 cm, weight approx. 7,5 kg (without sampling heads)
Specified tube/pipe length from sampling head to electronic unit of 10 m, tested up to 50 m
Operation with 90 to 100 mm nutrient plates
Fully automatic calibration of air flow with MAS-100 Regulus digital anemometer
Certificaten: Validation documents in accordance with ISO 14698 part 1 and part 2 Available with MAS®-100.