Individual pore size columns, PLgel
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PL1110-6530 PL1110-6540 PL1110-6550 PL1510-5520 PL1510-5540 PL1510-5525 PL1510-5530
HEWLPL1110-6530EA 2254 EUR
HEWLPL1110-6530 HEWLPL1110-6540 HEWLPL1110-6550 HEWLPL1510-5520 HEWLPL1510-5540 HEWLPL1510-5525 HEWLPL1510-5530
Individual pore size columns, PLgel
Kolommen voor chromatografie
High resolution over a specific MW range. Typical applications: Triglycerides, linear hydrocarbons, acrylates, rubbers. Compatible with organic solvents.
  • High speed and efficiency saves operator time
  • Maximise the resolution over a specific MW range
Individual pore size GPC columns offer the highest resolution within a narrow molecular weight range. The linear portion of the calibration curve, where the slope is at its shallowest, defines the MW region over which optimum resolution will be achieved.