Ion lenses for 7700/7800/7900 and 8800/8900
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G1833-65592 G1833-65593 G3280-01650 G3280-20618 G3280-20628 G3280-20633 G3280-20646 G3280-20653 G3280-20663 G3280-20664 G3280-60609 G3280-60612 G3280-60613 G3280-60615 G3280-67035 G3280-67037 G3280-67038 G3280-67039 G3280-67044 G3280-67045 G3280-67046 G8420-60700 G8400-67701
HEWLG1833-65592EA 602 EUR
HEWLG1833-65592 HEWLG1833-65593 HEWLG3280-01650 HEWLG3280-20618 HEWLG3280-20628 HEWLG3280-20633 HEWLG3280-20646 HEWLG3280-20653 HEWLG3280-20663 HEWLG3280-20664 HEWLG3280-60609 HEWLG3280-60612 HEWLG3280-60613 HEWLG3280-60615 HEWLG3280-67035 HEWLG3280-67037 HEWLG3280-67038 HEWLG3280-67039 HEWLG3280-67044 HEWLG3280-67045 HEWLG3280-67046 HEWLG8420-60700 HEWLG8400-67701
Ion lenses for 7700/7800/7900 and 8800/8900
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
High quality off-axis design ion lenses separate ions from photons and residual neutral material resulting in lower background noise in 7700/8800 ICP-MS systems.
  • High transmission off-axis or omega lens arrangement that separates the positively charged ions from the photons and neutral particles
  • Two types of extraction lens are available. The extraction lenses are placed just behind the skimmer cone and mounted on the skimmer base. They are used to extracts positive ions and accelerate them to the Einzel lens
  • The omega lens is a block of 5 lenses (omega
More Product Information
Order Now
Specificaties
Environmentally Preferable