Capillary and fitting kits
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5065-9947 G1316-68716
HEWL5065-9947EA 663 EUR
HEWL5065-9947 HEWLG1316-68716
Capillary and fitting kits
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These starter kits contain the most widely used capillary tubing, Swagelok connectors, and fittings in a variety of sizes.
  • Designed-in for highest chromatographic performance
  • High quality products for lowest instrument downtime and best total cost of ownership
  • OEM products fit tightest Agilent specifications for validated environments
We’ve also included our flexible stainless steel capillaries to help you make the best LC connections, regardless of equipment brand. Plus, as a special bonus, all multi-use kits (HEWL5065-9938, HEWL5065-9937, HEWL5065-9939) feature a FREE cybertool that puts more than 30 lab essentials at fingertips. For high-efficiency columns, it’s best to use narrow-diameter red tubing (Int.Ø 0,12 mm), instead of conventional green (Int.Ø 0,17 mm) tubing.