Coconut powder
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BIPC187-100G BIPC187-1KG BIPC187-500G
APOSBIPC187-100GEA 29.2 EUR
APOSBIPC187-100G APOSBIPC187-1KG APOSBIPC187-500G
Coconut powder
Foodservice benodigdheden
A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.