Heller's basal salt mixture
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMH393-10X1L PMH393-10L PMH393-50L
APOSPMH393-10X1LEA 0 EUR
APOSPMH393-10X1L APOSPMH393-10L APOSPMH393-50L
Heller's basal salt mixture
Celcultuurmedia
Contains macro/micronutrients as described by Heller, 1953 Plant tissue culture testedFull specification given in appendix.
  • MFCD00283413