Kao and Michayluk basal salt mixture
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMK413-10L PMK413-50L
APOSPMK413-10LEA 0 EUR
APOSPMK413-10L APOSPMK413-50L
Kao and Michayluk basal salt mixture
Celcultuurmedia
Contains macro/micronutrients required to initiate plant growthPlant tissue culture testedFull specification given in appendix.
  • MFCD00283578