Murashige and skoog modified basal salts (contains no N, P or K)
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMM407A-10X1L PMM407B-10L PMM407C-50L
APOSPMM407A-10X1LEA 35.3 EUR
APOSPMM407A-10X1L APOSPMM407B-10L APOSPMM407C-50L
Murashige and skoog modified basal salts (contains no N, P or K)
Celcultuurmedia
Contains macro/micronutrients as described by Murashige and Skoog, 1962. Modified by eliminating NH4NO3 and KNO3. Plant tissue culture tested. Full specification given in appendix.