Murashige and skoog Medium with vitamins and sucrose
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMM5501B-10L PMM5501C-50L PMM5501D-100L
APOSPMM5501B-10LEA 51.4 EUR
APOSPMM5501B-10L APOSPMM5501C-50L APOSPMM5501D-100L
Murashige and skoog Medium with vitamins and sucrose
Celcultuurmedia
Modified version of product number PMM519, contains 30 g/L D-Sucrose. Plant Tissue Culture Tested. Soluble in water.