Murashige and skoog modified basal medium (with kinetin)
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMM555-10L PMM555-50L
APOSPMM555-10LEA 0 EUR
APOSPMM555-10L APOSPMM555-50L
Murashige and skoog modified basal medium (with kinetin)
Celcultuurmedia
Contains macro/micronutrients as described by Murashige and Skoog, 1962. Modified with addition ofKinetin and NAA. Plant tissue culture testedFull specification given in appendix.
  • MFCD00240976