White's basal salt mixture
Leverancier: Apollo Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PMW898A-10X1L PMW898B-10L PMW898C-50L
APOSPMW898A-10X1LEA 0 EUR
APOSPMW898A-10X1L APOSPMW898B-10L APOSPMW898C-50L
White's basal salt mixture
Celcultuurmedia
With the macro/micronutrients as described by White, 1968. Full specification given in appendix.
  • MFCD00284888