ICP-OES torch injectors for PerkinElmer
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8003-0365 8003-0366 8003-0368 8003-0369 8003-0370 8003-0371 8003-0372 8003-0373 8003-0374 8003-0375 8003-0453 8003-0952
HEWL8003-0365EA 439 EUR
HEWL8003-0365 HEWL8003-0366 HEWL8003-0368 HEWL8003-0369 HEWL8003-0370 HEWL8003-0371 HEWL8003-0372 HEWL8003-0373 HEWL8003-0374 HEWL8003-0375 HEWL8003-0453 HEWL8003-0952
ICP-OES torch injectors for PerkinElmer
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
These torch injectors for PerkinElmer ICP-OES are available in a wide range of types. Choose from alumina or quartz in a range of sizes and acid resistance.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage