HPLC columns, InfinityLab Poroshell 120 EC-C18
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
693970-902K 695770-902K 695970-902K 650750-902K 690970-302K 690970-902K
HEWL693970-902KEA 2152 EUR
HEWL693970-902K HEWL695770-902K HEWL695970-902K HEWL650750-902K HEWL690970-302K HEWL690970-902K
HPLC columns, InfinityLab Poroshell 120 EC-C18
Kolommen voor chromatografie
Rugged, high-resolution separations up to five times faster than traditional 5 µm methods.
  • High efficiency columns for improved throughput
  • Excellent lot-to-lot reproducibility gives confidence in separation results
  • New HILIC chemistries offer a range of options for challenging polar compounds
InfinityLab Poroshell 120 columns provide exceptional efficiency and reliability, significantly boosting performance from all instruments, whether need improved HPLC performance, UHPLC performance at lower pressures or the highest UHPLC performance. The three particle diameters 1,9, 2,7 and 4 μm allow to choose the size that best fits separation needs and is most compatible with LC systems, and 12 different chemistries provide a range of selectivities that makes your method development fast and easy.