96-Well plates, VersaPlate™
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
75409025 7540801C 7540802C 75406050 75480060
VARI75409025EA 482 EUR
VARI75409025 VARI7540801C VARI7540802C VARI75406050 VARI75480060
96-Well plates, VersaPlate™
Microplaten Platen voor vastefase-extractie
VersaPlate allows you to process more samples in less time with less waste in a 96-well format.
  • Use only the number of wells you need for each run to save on sample preparation costs, while still enjoying all the benefits of automation
  • Increased well depth reduces the risk of overflow, and flangeless, beveled rims provide a tight fit. This helps avoid cross contamination
  • Scale up using the exact same product that was used with small scale development
Develop an SPE method faster, using liquid handling automation or multichannel pipettes, or use individual tubes to quickly work out method conditions and transfer to 96-well processing.
The Agilent VersaPlate allows you to process more samples in less time with less waste. In its 96-well format, tubes are removable, making this system easier to implement, simpler to use, and far more flexible than conventional 96-well formats. Whether you need to speed the processing of hundreds of samples, validate just a few samples, or develop a new method, you can customise the configuration to meet your needs.