Connectors, for GL threaded connections
Leverancier: Bohlender
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D584-18 D584-14 D583-25 D583-18 D583-14
201-0271EA 6 EUR
201-0271 BOHLD584-14 BOHLD583-25 BOHLD583-18 BOHLD583-14
Connectors, for GL threaded connections
Fittingen en connectoren
Connector made from PP with elastic sealing lip and FPM O-ring
  • Good chemical resistance
  • Temperature resistant from −20 to +110°C
  • Suitable for vacuum applications
For use with GL screw caps.